Anti-TERT Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A72989-100UL A72989-50UL
77847-213EA 521.4 CAD
77847-213 77847-214
Anti-TERT Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA:1:1000 to 1:2000, WB:1:200 to 1:1000, IHC:1:25 to 1:100

Type:
Antigen: TERT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR