Hemocyanin (from Keyhole Limpet (Megathura crenulata))
Supplier: MilliporeSigma
374817-1GM 374817-250MG 374817-50MG
CA80056-658EA 3652.43 CAD
CA80056-658 CA80056-660 CA80056-662
Hemocyanin (from Keyhole Limpet (Megathura crenulata))
Proteins and Peptides
Hemocyanin, Keyhole Limpet, Megathura crenulata, Glycerol, Hemocyanin, Keyhole Limpet, Megathura crenulata, Glycerol, is a large, multi-subunit, oxygen-carrying, metalloprotein that is used as a carrier protein in the production of antibodies.