Anti-HIST1H3A (tri-methyl-K9) Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A24962-100UL A24962-50UL
77739-899EA 553 CAD
77739-899 77857-177
Anti-HIST1H3A (tri-methyl-K9) Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IF: 1:50 to 1:200

Type:
Antigen: HIST1H3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR