Anti-HIST1H2BC Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A24906-100UL A24906-50UL
77739-857EA 553 CAD
77739-857 77857-121
Anti-HIST1H2BC Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:100 to 1:1000, IHC: 1:10 to 1:100, IF: 1:1 to 1:10

Type:
Antigen: HIST1H2BC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR