Anti-INPP5B Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A26345-100UG A26345-50UG
77741-103EA 521.4 CAD
77741-103 77858-127
Anti-INPP5B Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: INPP5B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR