Anti-HIST1H1C Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A24687-100UL A24687-50UL
77739-725EA 553 CAD
77739-725 77856-902
Anti-HIST1H1C Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:10-1:100

Type:
Antigen: HIST1H1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR