Anti-HIST1H3A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A25029-100UL A25029-50UL
77739-942EA 553 CAD
77739-942 77857-244
Anti-HIST1H3A Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:100 to 1:1000, IHC: 1:10 to 1:100

Type:
Antigen: HIST1H3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR