Anti-NRG2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A30061-100UG A30061-50UG
77745-035EA 521.4 CAD
77745-035 77745-036
Anti-NRG2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: NRG2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol NRG2
Environmentally Preferable
Antigen synonyms Divergent of neuregulin 1|Divergent of neuregulin-1|Don 1|DON-1|Neural- and thymus-derived activator for ERBB kinases|Neuregulin 2|Neuregulin-2|NRG-2|NRG2_HUMAN|NTAK|Pro Pro-NRG2
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID O14511
Isotype IgG
ELISA Yes
Immunogen Pro-neuregulin-2, membrane-bound isoform protein (112-405AA)
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR