Anti-HK1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A25243-100UG A25243-50UG
77740-093EA 521.4 CAD
77740-093 77857-451
Anti-HK1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: HK1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR