Anti-HIST1H4A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A25195-100UL A25195-50UL
77740-055EA 553 CAD
77740-055 77857-408
Anti-HIST1H4A Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:50 to 1:500, IHC:1:10 to 1:100

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR