Anti-NRARP Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A30038-100UG A30038-50UG
77744-995EA 521.4 CAD
77744-995 77744-996
Anti-NRARP Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: NRARP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol NRARP
Environmentally Preferable
Antigen synonyms Notch regulated ankyrin repeat containing protein|NOTCH regulated ankyrin repeat protein|MGC61598|Notch-regulated ankyrin repeat-containing protein|NRARP_HUMAN
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID Q7Z6K4
Isotype IgG
ELISA Yes
Immunogen Notch-regulated ankyrin repeat-containing protein (1-114AA)
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR