Anti-ILK Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A24117-100UG A24117-50UG
77739-208EA 521.4 CAD
77739-208 77856-579
Anti-ILK Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: ILK
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR