Anti-GPHN Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A23642-100UG A23642-50UG
77738-749EA 521.4 CAD
77738-749 77856-311
Anti-GPHN Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: GPHN
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR