Anti-MEN1 Rabbit Recombinant polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A28435-100UG A28435-50UG
77743-006EA 521.4 CAD
77743-006 77859-662
Anti-MEN1 Rabbit Recombinant polyclonal Antibody (Biotinylated)
Antibodies
0

Type:
Antigen: MEN1
Clonality: Recombinant polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR