Anti-FLT3 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A22100-100UL A22100-50UL
77737-298EA 521.4 CAD
77737-298 77855-476
Anti-FLT3 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:1000 to 1:5000

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR