Anti-H2AFZ (mono-methyl-K7) Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A24297-100UL A24297-50UL
77739-373EA 553 CAD
77739-373 77856-693
Anti-H2AFZ (mono-methyl-K7) Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ICC:1:20 to 1:200, IF: 1:50 to 1:200

Type:
Antigen: H2AFZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR