Anti-HIST1H4A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A25168-100UL A25168-50UL
77740-040EA 553 CAD
77740-040 77857-381
Anti-HIST1H4A Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:1 to 1:10

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR