Anti-FMOD Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A22143-100UG A22143-50UG
77737-340EA 521.4 CAD
77737-340 77855-502
Anti-FMOD Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: FMOD
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR