Anti-OGT Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A30301-50UG A30301-100UG
77745-417EA 355.5 CAD
77745-417 77745-416
Anti-OGT Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: OGT
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR