19062

Supplier: AFG Bioscience
A29223-100UG A29223-50UG
77743-737EA 521.4 CAD
77743-737 77860-201
19062
Antibodies
Recommended dilution: IHC: 1:500 to 1:1000

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR