Anti-HIST1H4A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A25221-100UL A25221-50UL
77740-074EA 553 CAD
77740-074 77857-434
Anti-HIST1H4A Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ICC: 1:20 to 1:200, IF: 1:50 to 1:200

Type:
Antigen: HIST1H4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR