Anti-MAPK14 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: AFG Bioscience
A28144-100UG A28144-50UG
77742-742EA 521.4 CAD
77742-742 77859-458
Anti-MAPK14 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type:
Antigen: MAPK14
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR