Anti-NOS1AP Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A29842-50UG A29842-100UG
77744-650EA 355.5 CAD
77744-650 77744-649
Anti-NOS1AP Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: NOS1AP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol NOS1AP
Environmentally Preferable
Antigen synonyms C terminal PDZ ligand of neuronal nitric oxide synthase protein|Nitric oxide synthase 1 (neuronal) adaptor protein|C terminal PDZ domain ligand of neuronal nitric oxide synthase|C-terminal PDZ ligand of neuronal nitric oxide synthase protein|C terminal PDZ domain ligand of neuronal nitric oxide synthase (CAPON)|Carboxyl-terminal PDZ ligand of neuronal nitric oxide synthase protein|Nitric oxide synthase 1 adaptor protein|6330408P19Rik|MGC138500|Carboxyl terminal PDZ ligand of neuronal nitric oxide synthase protein|Ligand of neuronal nitric oxide synthase with carboxyl terminal PDZ domain|CAPON|CAPON_HUMAN
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID O75052
Isotype IgG
ELISA Yes
Immunogen Carboxyl-terminal PDZ ligand of neuronal nitric oxide synthase protein (176-288AA)
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR