14642

Supplier: AFG Bioscience
A24761-100UL A24761-50UL
77739-768EA 553 CAD
77739-768 77856-976
14642
Antibodies
Recommended dilution: IF: 1:50 to 1:200

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR