Anti-GSC Rabbit Recombinant polyclonal Antibody

Supplier: AFG Bioscience
A23989-100UG A23989-50UG
77739-088EA 521.4 CAD
77739-088 77856-497
Anti-GSC Rabbit Recombinant polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:200-1:500

Type:
Antigen: GSC
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR