Anti-INPP1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A26332-100UG A26332-50UG
77741-090EA 521.4 CAD
77741-090 77858-116
Anti-INPP1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR