19021

Supplier: AFG Bioscience
A29177-100UG A29177-50UG
77743-697EA 521.4 CAD
77743-697 77860-171
19021
Antibodies
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR