Anti-NTHL1 Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A30090-50UG A30090-100UG
77745-079EA 355.5 CAD
77745-079 77745-078
Anti-NTHL1 Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: NTHL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR