Anti-EMP1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A20557-100UG A20557-50UG
77735-864EA 521.4 CAD
77735-864 77854-703
Anti-EMP1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: EMP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol EMP1
Environmentally Preferable
Antigen synonyms CL-20|EMP-1|EMP1_HUMAN|Epithelial membrane protein 1|Protein B4B|TMP|Tumor-associated membrane protein
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID P54849
Isotype IgG
ELISA Yes
Immunogen Recombinant Human Epithelial membrane protein 1 protein (22-66AA)
Form Liquid
Purification Antigen affinity purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR