Anti-HIST1H2AG Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A24788-100UL A24788-50UL
77739-789EA 553 CAD
77739-789 77857-003
Anti-HIST1H2AG Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:50 to 1:500, IHC:1:20 to 1:200, IF:1:1 to 1:10

Type:
Antigen: HIST1H2AG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR