20311

Supplier: AFG Bioscience
A30489-100UL A30489-50UL
77745-743EA 521.4 CAD
77745-743 77745-744
20311
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR