Anti-OTC Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A30474-50UG A30474-100UG
77745-718EA 355.5 CAD
77745-718 77745-717
Anti-OTC Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:200 to 1:500

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR