Anti-HIST1H2AG Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A24807-100UL A24807-50UL
77739-800EA 553 CAD
77739-800 77857-022
Anti-HIST1H2AG Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:200-1:2000, IHC:1:20-1:200

Type:
Antigen: HIST1H2AG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR