Anti-IGHD Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A25991-100UG A25991-50UG
77740-771EA 521.4 CAD
77740-771 77857-947
Anti-IGHD Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: IGHD
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR