Anti-APC Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A25514-100UL A25514-50UL
77740-341EA 521.4 CAD
77740-341 77857-620
Anti-APC Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA:1:1000 to 1:2000, IHC:1:25 to 1:100

Type:
Antigen: APC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR