Anti-MMP14 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A28687-100UL A28687-50UL
77743-243EA 521.4 CAD
77743-243 77859-841
Anti-MMP14 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR