Anti-GNAQ Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A23425-100UG A23425-50UG
77738-542EA 521.4 CAD
77738-542 77856-178
Anti-GNAQ Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20 to 1:200, IF:1:50 to 1:200

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR