Anti-RPS18 Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A30394-100UG A30394-50UG
77745-580EA 521.4 CAD
77745-580 77745-581
Anti-RPS18 Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: RPS18
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR