Anti-ESM1 Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A20900-100UG A20900-50UG
77736-179EA 521.4 CAD
77736-179 77854-922
Anti-ESM1 Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
0

Type:
Antigen: ESM1
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR