Anti-GRP Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A23980-100UG A23980-50UG
77739-079EA 521.4 CAD
77739-079 77856-492
Anti-GRP Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: GRP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR