Anti-GRP Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A30441-50UL A30441-100UL
77745-672EA 355.5 CAD
77745-672 77745-671
Anti-GRP Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000

Type:
Antigen: GRP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR