Anti-HIST1H4A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A25157-100UL A25157-50UL
77740-034EA 553 CAD
77740-034 77857-370
Anti-HIST1H4A Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IF: 1:1 to 1:10

Type:
Antigen: HIST1H4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR