Anti-NDUFB8 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A29548-100UG A29548-50UG
77744-202EA 521.4 CAD
77744-202 77744-203
Anti-NDUFB8 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: NDUFB8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol NDUFB8
Environmentally Preferable
Antigen synonyms ASHI|CI-ASHI|Complex I ASHI subunit|Complex I-ASHI|mitochondrial|NADH dehydrogenase (ubiquinone) 1 beta subcomplex 8 19kDa|NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 8|NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 8 mitochondrial|NADH-ubiquinone oxidoreductase ASHI subunit|NDUB8_HUMAN
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID O95169
Isotype IgG
ELISA Yes
Immunogen NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 8, mitochondrial protein (29-132AA)
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
Storage temperature Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR