Anti-HMBS Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A25289-100UG A25289-50UG
77740-133EA 521.4 CAD
77740-133 77857-473
Anti-HMBS Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: HMBS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR