Anti-HIST1H3A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A25044-100UL A25044-50UL
77739-955EA 553 CAD
77739-955 77857-259
Anti-HIST1H3A Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:200 to 1:2000, IHC: 1:20 to 1:200

Type:
Antigen: HIST1H3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR