Anti-H2AFJ Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A24277-100UG A24277-50UG
77739-357EA 521.4 CAD
77739-357 77856-677
Anti-H2AFJ Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
0

Type:
Antigen: H2AFJ
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR