Anti-HIST1H2BC Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A24907-100UL A24907-50UL
77739-858EA 553 CAD
77739-858 77857-122
Anti-HIST1H2BC Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:10 to 1:100

Type:
Antigen: HIST1H2BC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR