Anti-HIST1H4A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A25223-100UL A25223-50UL
77740-075EA 553 CAD
77740-075 77857-436
Anti-HIST1H4A Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:200 to 1:2000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200, IP: 1:200 to 1:2000

Type:
Antigen: HIST1H4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR