Anti-ERBB3 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A20798-100UL A20798-50UL
77736-086EA 521.4 CAD
77736-086 77854-873
Anti-ERBB3 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20 to 1:200

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR