Anti-HIST1H4A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A25166-100UL A25166-50UL
77740-038EA 553 CAD
77740-038 77857-379
Anti-HIST1H4A Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:100 to 1:1000, IF: 1:1 to 1:10

Type:
Antigen: HIST1H4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR